Learn More
Enzo Life Sciences Clindamycin. HCl (100mg). CAS: 21462-39-5
SDP
Catalog No. 502010228
Encompass_Preferred
Active against Gram-positive bacteria. Clindamycin binds to the 50S subunit of bacterial ribosomes and suppresses protein synthesis. Alternative name: U-21251F. Purity: ≥95% (HPLC). Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-201-0228 Supplier Enzo Life Sciences Supplier No. BMLA1900100
Active against Gram-positive bacteria. Clindamycin binds to the 50S subunit of bacterial ribosomes and suppresses protein synthesis. Alternative name: U-21251F. Purity: ≥95% (HPLC). Solubility: Soluble in water. Long Term Storage: +4°C.