Learn More
Selleck Chemical LLC Clindamycin HCl S2457-1g
SDP
Catalog No. 503112613
Encompass_Preferred
Clindamycin HCl inhibits protein synthesis by acting on the 50S ribosomal used for the treatment of bacterial infections
Catalog No. 50-311-2613 Supplier Selleck Chemical LLC Supplier No. S24571G
Clindamycin HCl inhibits protein synthesis by acting on the 50S ribosomal used for the treatment of bacterial infections