Learn More
Apexbio Technology LLC Clindamycin HCl 21462-39-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063390
Encompass_Preferred
Clindamycin hydrochloride is a semi-synthetic antibiotic of the lincosamide family exerting antimicrobial activity by directly binding to the bacterial ribosome 50S subunit and subsequently inhibiting protein synthesis Its mechanism specifically disrupts the elongation process during peptide chain formation This compound is frequently utilized in microbiological and biochemical research protocols to study bacterial translation mechanisms evaluate susceptibility profiles in antimicrobial resistance assays and investigate bacterial response pathways related to protein metabolism The reported IC50 values for Clindamycin hydrochloride typically range between 0 02 0 5 g/mL depending on bacterial strains tested making it suitable for in vitro assessments of bacterial resistance phenotypes and antimicrobial susceptibility
Catalog No. 50-000-63390 Supplier Apexbio Technology LLC Supplier No. B19135.1
Clindamycin hydrochloride is a semi-synthetic antibiotic of the lincosamide family exerting antimicrobial activity by directly binding to the bacterial ribosome 50S subunit and subsequently inhibiting protein synthesis Its mechanism specifically disrupts the elongation process during peptide chain formation This compound is frequently utilized in microbiological and biochemical research protocols to study bacterial translation mechanisms evaluate susceptibility profiles in antimicrobial resistance assays and investigate bacterial response pathways related to protein metabolism The reported IC50 values for Clindamycin hydrochloride typically range between 0 02 0 5 g/mL depending on bacterial strains tested making it suitable for in vitro assessments of bacterial resistance phenotypes and antimicrobial susceptibility