Learn More
Selleck Chemical LLC Clindamycin HCl 50mg 21462-39-5
SDP
Catalog No. 505113
Encompass_Preferred
Clindamycin HCl inhibits protein synthesis by acting on the 50S ribosomal, used for the treatment of bacterial infections. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-511-3 Supplier Selleck Chemical LLC Supplier No. S245750MG
Clindamycin HCl inhibits protein synthesis by acting on the 50S ribosomal, used for the treatment of bacterial infections. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.