Learn More
Bioworld Clindamycin HCl, 1 g
Catalog No. 50255720
Encompass_Preferred
Clindamycin, HCl Lincosamide antibiotic active against Gram-positive bacteria.Synonym: U-21251F; Cleocin; ClinimycinA suppressor of protein synthesis in bacteriaFor in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 647 °C at 760 mmHg (Predicted)Melting Point: 141 °CRTECS Number: GF2275000
Catalog No. 50-255-720 Supplier Bioworld Supplier No. 403100241
Clindamycin, HCl Lincosamide antibiotic active against Gram-positive bacteria.Synonym: U-21251F; Cleocin; ClinimycinA suppressor of protein synthesis in bacteriaFor in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 647 °C at 760 mmHg (Predicted)Melting Point: 141 °CRTECS Number: GF2275000