Learn More
Medchemexpress LLC Clindamycin (hydrochloride) | 21462-39-5 | 99.9% | 1 G
SDP
Catalog No. 5000409239
Encompass_Preferred
Clindamycin (hydrochloride) is a semisynthetic lincosamide antibiotic that inhibits protein synthesis by acting on the 50S ribosomal subunit. It is for research use only.
  • Semisynthetic lincosamide antibiotic
  • Inhibits protein synthesis by acting on the 50S ribosomal subunit
  • White to off-white solid
  • Purity of 99.9%
Catalog No. 50-004-09239 Supplier Medchemexpress LLC Supplier No. HYB0408A1G
Clindamycin (hydrochloride) is a semisynthetic lincosamide antibiotic that inhibits protein synthesis by acting on the 50S ribosomal subunit. It is for research use only.
  • Semisynthetic lincosamide antibiotic
  • Inhibits protein synthesis by acting on the 50S ribosomal subunit
  • White to off-white solid
  • Purity of 99.9%