Learn More
Medchemexpress LLC Clindamycin hydrochloride | 21462-39-5 | 99.9% | 100 MG
SDP
Catalog No. 5000406781
Encompass_Preferred
Clindamycin hydrochloride is a semisynthetic lincosamide antibiotic, which inhibits protein synthesis by acting on the 50S ribosomal.
  • Semisynthetic lincosamide antibiotic
  • Appears as a white to off-white solid
  • High solubility in DMSO and H2O (≥ 100 mg/mL)
  • Recommended storage at 4°C for solid, -80°C for long-term in solvent
Catalog No. 50-004-06781 Supplier Medchemexpress LLC Supplier No. HYB0408A100MG
Clindamycin hydrochloride is a semisynthetic lincosamide antibiotic, which inhibits protein synthesis by acting on the 50S ribosomal.
  • Semisynthetic lincosamide antibiotic
  • Appears as a white to off-white solid
  • High solubility in DMSO and H2O (≥ 100 mg/mL)
  • Recommended storage at 4°C for solid, -80°C for long-term in solvent