Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clindamycin hydrochloride | 21462-39-5 | 99.9% | 5 G
SDP

Catalog No. 5000409173
Encompass_Preferred

Clindamycin hydrochloride is a semisynthetic lincosamide antibiotic that inhibits protein synthesis by acting on the 50S ribosomal subunit.

  • Inhibits protein synthesis
  • Semisynthetic lincosamide antibiotic
  • High purity: 99.9%
  • Solid appearance
  • White to off-white color
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture

Catalog No. 50-004-09173 Supplier Medchemexpress LLC Supplier No. HYB0408A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clindamycin hydrochloride is a semisynthetic lincosamide antibiotic that inhibits protein synthesis by acting on the 50S ribosomal subunit.

  • Inhibits protein synthesis
  • Semisynthetic lincosamide antibiotic
  • High purity: 99.9%
  • Solid appearance
  • White to off-white color
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.