Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clindamycin Hydrochloride, G-Biosciences
SCI_ED

Catalog No. S100937
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S100937 25 g
S000031 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S100937 Supplier G-Biosciences Supplier No. RC979
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clindamycin is a lincosamide antibiotic primarily targeting gram-positive bacteria. It is also against anaerobic bacteria and protozoans such as Toxoplasma species.

Clindamycin binds the 50s ribosomal subunit preventing ribosomal translocation, resulting in inhibition of protein synthesis. It has been shown to exhibit synergistic properties when combined with vancomycin.

Chemical Identifiers

CAS 21462-39-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 25 g
Percent Purity 98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.