Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clindamycin Hydrochloride USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048059 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-059 100 g
18-048-060 500 g
18-048-078 5 g
18-048-079 25 g
18-048-080 1 kg
18-048-104 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-048-059 Supplier Medisca Supplier No. 000505
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clindamycin Hydrochloride USP is a white or practically white, crystalline powder. It is odorless to having a faint mercaptan-like odor. Stable in the presence of air and light.

Chemical Identifiers

CAS 21462-39-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White
Quantity 100 g
Percent Purity ≥ 800 μg
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.