Learn More
U.S. Pharmacopeia Clindamycin Palmitate Hydrochloride, 25507-04-4, MFCD09752824, 500mg
Catalog No. NC1401631
Encompass
Molecular formula C34H63ClN2O6S . HCl, Molecular weight 699.85, Melting Point 334.4 - 352.4 °F (168 - 178 °C)
Catalog No. NC1401631 Supplier U.S. Pharmacopeia Supplier No. 1137005
Molecular formula C34H63ClN2O6S . HCl, Molecular weight 699.85, Melting Point 334.4 - 352.4 °F (168 - 178 °C)