Learn More
Medchemexpress LLC Clindamycin palmitate hydrochloride | 25507-04-4 | 98.2% | 1 ML
SDP
Catalog No. 5000413426
Encompass_Preferred
Clindamycin palmitate hydrochloride is a hydrochloride salt of the ester of clindamycin and palmitic acid. It functions as an antibacterial agent. While it is inactive in vitro, it rapidly hydrolyzes in vivo to become the antibacterially active clindamycin.
  • Functions as an antibacterial agent.
  • Inactive in vitro, but rapidly converts to active clindamycin in vivo.
  • Available in various forms, including solid and solution.
  • Strict storage conditions ensure product stability and efficacy.
Catalog No. 50-004-13426 Supplier Medchemexpress LLC Supplier No. HYB145410MM/1ML
Clindamycin palmitate hydrochloride is a hydrochloride salt of the ester of clindamycin and palmitic acid. It functions as an antibacterial agent. While it is inactive in vitro, it rapidly hydrolyzes in vivo to become the antibacterially active clindamycin.
  • Functions as an antibacterial agent.
  • Inactive in vitro, but rapidly converts to active clindamycin in vivo.
  • Available in various forms, including solid and solution.
  • Strict storage conditions ensure product stability and efficacy.