Learn More
Cayman Chemical Clindamycin Palmitate (hydrochloride), Purity: ≥95%, CAS: 25507-04-4, 100 mg
SDP
Catalog No. 502940307
Encompass_Preferred
A water-soluble prodrug form of clindamycin; undergoes rapid hydrolysis in vivo to release active clindamycin
Catalog No. 50-294-0307 Supplier Cayman Chemical Supplier No. 29102100 MG
A water-soluble prodrug form of clindamycin; undergoes rapid hydrolysis in vivo to release active clindamycin