Learn More
Cayman Chemical Clindamycin Palmitate (hydrochloride), Purity: ≥95%, CAS: 25507-04-4, 250 mg
SDP
Catalog No. 502938174
Encompass_Preferred
A water-soluble prodrug form of clindamycin; undergoes rapid hydrolysis in vivo to release active clindamycin
Catalog No. 50-293-8174 Supplier Cayman Chemical Supplier No. 29102250 MG
A water-soluble prodrug form of clindamycin; undergoes rapid hydrolysis in vivo to release active clindamycin