Learn More
Medchemexpress LLC Clindamycin phosphate | 24729-96-2 | MFCD11982855 | 98.0% | 1 G
SDP
Catalog No. 5000412459
Encompass_Preferred
Clindamycin phosphate is a broad-spectrum bacteriostatic lincosamide antibiotic. It acts as a proagent, converting in vivo to the active form, Clindamycin, through phosphatase ester hydrolysis. This compound is used in research for conditions such as acne and bacterial vaginosis, demonstrating no cytotoxicity. Its antimicrobial properties are enhanced when combined with platelet rich fibrin (PRF).
  • Acts as a broad-spectrum bacteriostatic lincosamide antibiotic.
  • Converts to active Clindamycin in vivo.
  • Shows no cytotoxicity.
  • Enhances antimicrobial properties when combined with platelet rich fibrin.
  • Suitable for research and analytical applications.
Catalog No. 50-004-12459 Supplier Medchemexpress LLC Supplier No. HYB10641G
Clindamycin phosphate is a broad-spectrum bacteriostatic lincosamide antibiotic. It acts as a proagent, converting in vivo to the active form, Clindamycin, through phosphatase ester hydrolysis. This compound is used in research for conditions such as acne and bacterial vaginosis, demonstrating no cytotoxicity. Its antimicrobial properties are enhanced when combined with platelet rich fibrin (PRF).
  • Acts as a broad-spectrum bacteriostatic lincosamide antibiotic.
  • Converts to active Clindamycin in vivo.
  • Shows no cytotoxicity.
  • Enhances antimicrobial properties when combined with platelet rich fibrin.
  • Suitable for research and analytical applications.