Learn More
Medchemexpress LLC Clindamycin phosphate | 24729-96-2 | MFCD11982855 | 99.5% | 500 MG
SDP
Catalog No. 5000411810
Encompass_Preferred
Clindamycin phosphate is a broad-spectrum bacteriostatic lincosamide antibiotic. It is a proagent of Clindamycin and has no antimicrobial activity in vitro, but is rapidly converted in vivo to the active parent agent, Clindamycin, by phosphatase ester hydrolysis. It can be used for researching acne and bacterial vaginosis. Clindamycin phosphate has no cytotoxicity.
  • Broad-spectrum bacteriostatic lincosamide antibiotic
  • Proagent of Clindamycin
  • Rapidly converted in vivo to active Clindamycin
  • Used for researching acne and bacterial vaginosis
  • Exhibits no cytotoxicity
  • Antimicrobial properties enhanced when combined with platelet rich fibrin (PRF)
Catalog No. 50-004-11810 Supplier Medchemexpress LLC Supplier No. HYB1064500MG
Clindamycin phosphate is a broad-spectrum bacteriostatic lincosamide antibiotic. It is a proagent of Clindamycin and has no antimicrobial activity in vitro, but is rapidly converted in vivo to the active parent agent, Clindamycin, by phosphatase ester hydrolysis. It can be used for researching acne and bacterial vaginosis. Clindamycin phosphate has no cytotoxicity.
  • Broad-spectrum bacteriostatic lincosamide antibiotic
  • Proagent of Clindamycin
  • Rapidly converted in vivo to active Clindamycin
  • Used for researching acne and bacterial vaginosis
  • Exhibits no cytotoxicity
  • Antimicrobial properties enhanced when combined with platelet rich fibrin (PRF)