Learn More
Selleck Chemical LLC Clindamycin Phosphate S2048-50mg
SDP
Catalog No. 503110370
Encompass_Preferred
Clindamycin Phosphate (NSC 618653) is a lincosamide antibiotic for Plasmodium falciparum with IC50 of 12 nM
Catalog No. 50-311-0370 Supplier Selleck Chemical LLC Supplier No. S204850MG
Clindamycin Phosphate (NSC 618653) is a lincosamide antibiotic for Plasmodium falciparum with IC50 of 12 nM