Learn More
Cayman Chemical ClIndamycIn Phosphate 50g
SDP
Catalog No. 502913653
Encompass_Preferred
A lincosamide antibiotic; active against P. acnes in vitro (MIC = 0.02 UG/ml); topical administration reduces the number of bacteria at the burn site in a mouse model of P. acnes burn infection at 1% w/w
Catalog No. 50-291-3653 Supplier Cayman Chemical Supplier No. 3561150 G
A lincosamide antibiotic; active against P. acnes in vitro (MIC = 0.02 UG/ml); topical administration reduces the number of bacteria at the burn site in a mouse model of P. acnes burn infection at 1% w/w