Learn More
Cayman Chemical Clindamycin Phosphate, Purity: ≥98%, CAS: 24729-96-2, 5 g
SDP
Catalog No. 502913957
Encompass_Preferred
A lincosamide antibiotic; active against P. acnes in vitro (MIC = 0.02 µg/ml); topical administration reduces the number of bacteria at the burn site in a mouse model of P. acnes burn infection at 1% w/w
Catalog No. 50-291-3957 Supplier Cayman Chemical Supplier No. 356115 G
A lincosamide antibiotic; active against P. acnes in vitro (MIC = 0.02 µg/ml); topical administration reduces the number of bacteria at the burn site in a mouse model of P. acnes burn infection at 1% w/w