Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys TruLab P is a lyophilized 'pathological' (high) multi-control for clinical chemistry (20x5mL)
Catalog No. NC2874320
Encompass_Preferred
DiaSys TruLab P is a lyophilized 'pathologic' (high) multi-control for clinical chemistry. For veterinary or research use only. Contains: Acetaminophen, ALAT, Albumin, Alkaline phosphatase, α-Amylase, Apolipoprotein A, Apolipoprotein B, ASAT, Bilirubin Direct and Total, Calcium, Carbamazepine, Chloride, Cholesterol, Cholinesterase, CK-NAC, CK-MB, Copper, Cortisol, Creatinine, Digoxin, DL-Phenylalanine, Free Cholesterol, Free Glycerol, Gamma-GT, Gentamycin, GLDH, Glucose, α-HBDH, HDL-Cholesterol, β-Hydroxybutyrate, IgA, IgE, IgG, IgM, Iron, Lactate, LDH, LDL-Cholesterol, Lipase, Lithium, Magnesium, Pancreatic Amylase, Phenytoin, Phospholipids, Phosphorus, Potassium, Salicylate, Sodium, Theophyllin, Tobramycin, Total Protein, Total Bile Acids, Transferrin, Triglycerides, TSH, Urea, Uric Acid, Zinc. DiaSys TruLab P must be stored refrigerated. Once reconstituted, DiaSys TruLab P is stable for 30 days when kept frozen.
This item is not returnable. View return policy
Catalog No. NC2874320 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 590509911062
DiaSys TruLab P is a lyophilized 'pathologic' (high) multi-control for clinical chemistry. For veterinary or research use only. Contains: Acetaminophen, ALAT, Albumin, Alkaline phosphatase, α-Amylase, Apolipoprotein A, Apolipoprotein B, ASAT, Bilirubin Direct and Total, Calcium, Carbamazepine, Chloride, Cholesterol, Cholinesterase, CK-NAC, CK-MB, Copper, Cortisol, Creatinine, Digoxin, DL-Phenylalanine, Free Cholesterol, Free Glycerol, Gamma-GT, Gentamycin, GLDH, Glucose, α-HBDH, HDL-Cholesterol, β-Hydroxybutyrate, IgA, IgE, IgG, IgM, Iron, Lactate, LDH, LDL-Cholesterol, Lipase, Lithium, Magnesium, Pancreatic Amylase, Phenytoin, Phospholipids, Phosphorus, Potassium, Salicylate, Sodium, Theophyllin, Tobramycin, Total Protein, Total Bile Acids, Transferrin, Triglycerides, TSH, Urea, Uric Acid, Zinc. DiaSys TruLab P must be stored refrigerated. Once reconstituted, DiaSys TruLab P is stable for 30 days when kept frozen.