Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC The DiaSys respons910VET is a benchtop clinical chemistry analyzer with open channels, small sample mode for <5 µL dead volumes, 150 tests/h throughput, ideal for research with mice and small samples.
Catalog No. NC2874352
Encompass_Preferred
The DiaSys respons910VET is the dedicated solution for veterinary or research in vitro-diagnostics - with over 30 parameters (ALAT (ALT/GPT), Albumin, Alkaline Phosphatase, Amylase, ASAT (AST/GOT), Bicarbonate, Bilirubin Auto Total, Bilirubin Auto Direct, BUN (=Urea), Calcium, Chloride, Cholesterol, CK-NAC, Creatinine, Gamma-GT, Glucose, ß-Hydroxybutyrate, Iron, Lactate, LDH, Lipase, Magnesium, NEFA, Phosphate, Potassium, Sodium, T4, Total Protein, Triglycerides, UIBC, Urea (=BUN), Uric Acid), plus unlimited open channels to address your testing needs. It is the perfect fit for laboratories with small to mid-range throughput. It offers full flexibility, from single test runs to full panel testing. Assays are adaptable to your needs. DiaSys Diagnostic Systems is the world leader in the development of liquid, ready to use products with extended room temperature stability. Extended linearity of our assays nearly eliminates repeat testing due to sample results being out of range.
Catalog No. NC2874352 Supplier DIASYS DIAGNOSTIC SYSTEMS USA LLC Supplier No. 960500
Add to Cart
Add to Cart
Request a Quote
The DiaSys respons910VET is the dedicated solution for veterinary or research in vitro-diagnostics - with over 30 parameters (ALAT (ALT/GPT), Albumin, Alkaline Phosphatase, Amylase, ASAT (AST/GOT), Bicarbonate, Bilirubin Auto Total, Bilirubin Auto Direct, BUN (=Urea), Calcium, Chloride, Cholesterol, CK-NAC, Creatinine, Gamma-GT, Glucose, ß-Hydroxybutyrate, Iron, Lactate, LDH, Lipase, Magnesium, NEFA, Phosphate, Potassium, Sodium, T4, Total Protein, Triglycerides, UIBC, Urea (=BUN), Uric Acid), plus unlimited open channels to address your testing needs. It is the perfect fit for laboratories with small to mid-range throughput. It offers full flexibility, from single test runs to full panel testing. Assays are adaptable to your needs. DiaSys Diagnostic Systems is the world leader in the development of liquid, ready to use products with extended room temperature stability. Extended linearity of our assays nearly eliminates repeat testing due to sample results being out of range.