Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Clinical Lab Mat
GREENER_CHOICE

Catalog No. 22267279 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
12 x 14.9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-267-279 12 x 14.9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-267-279
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fisherbrand™ Clinical Lab Mat safely facilitates workflow processes with organization and easy clean up, while keeping benchtops clean and safe from stains or spills.

  • Small footprint
  • Accommodates most work spaces
  • Helps facilitate workflow processes, such as spinning blood tubes in centrifuge workstations
  • Gridded surface for tube staging, preventing movement of small objects and securing items such as reservoirs, pipette tip boxes, assay plates, etc
  • Gridded surface enables tubes to be easily marked and identified
  • Personal size is easy to transport and allows users to easily create and protect temporary work areas
  • Reversible
  • Side One: combination gridded and smooth surface
  • Side Two: smooth surface only, two color options available
  • Lip around the edge can hold approx. 250 mL of liquid

Specifications

Product Type Clinical Lab Mat
Material Silicone
Absorbancy Non-absorbent
Length (English) 12 in.
Length (Metric) 30.5 cm
Width (English) 14.9 in.
Width (Metric) 37.9 cm
Dimensions (L x W) 12 x 14.9 in.
Backing Material Silicone
Color Blue
Disposable Reusable
Thickness 0.1 in.
Temperature (English) Max. 392°F
Temperature (Metric) Max 200°C
Quantity 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.