Learn More
Motic™ Clinical Package: PA43BIO Trino (Pathology) Upright Microscope w/ Moticam S6 and Image-Pro Life Science (2D) Software
Catalog No. 01673124 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Trinocular, Tilting
Catalog No. Head Style
01-673-124 Trinocular, Tilting
1 options
Catalog No. 01-673-124 Supplier Motic™ Supplier No. MBPA43PS6MC1
Add to Cart
Add to Cart
Request a Quote
This Clinical Package includes PA43BIO (Pathology) Trinocular Upright Microscope, Moticam S6 USB Camera, Image-Pro - Life Science Package for Motic (2D), 0.5X C-Mount camera adapter
  • Microscope: PA43BIO (Pathology) Trinocular Upright Microscope The PA43BIO (Pathology) Trinocular is a perfect workhorse for pathologist. This microscope has a simple design and is very ease to use, but also does not compromise on quality and ergonomic features.
  • It is a microscope meant to be used for long periods of time, with the highest quality of experience. The modularity of the PA43 microscopes offers you flexibility in choosing a configuration that suits your usage. It has a strong selection of contrast accessories that are available and easily upgradable to meet different requirements. Digital Camera: Moticam S6 USB Camera The Hi-Res Partner - Moticam S6 is designed for those application fields which need higher resolution.
  • Presentations to a greater audience through video projector and reports in hardcopy are performed with ease. With 6 million pixels and a fast data readout, the performance is remarkable! Software: Image-Pro - Life Science Package for Motic (2D) Transform your life sciences research with the Image-Pro - Life Science Package for Motic, developed in partnership with Media Cybernetics.
  • This versatile solution combines advanced Machine Learning technology with a comprehensive suite of protocols to provide precise segmentation, classification, and automated measurements from 2D images. Perfect for a wide range of life science applications, this package enhances your analytical capabilities with efficiency and accuracy.
  • Key Features:
  • Machine Learning-Powered Analysis Leverage sophisticated Machine Learning algorithms for accurate segmentation and classification, ensuring automated and reliable measurements from 2D images.
  • Cell Biology Protocol Collection Optimize your research with an extensive suite of protocols for analyzing complex cell biology-based measurements, including Cell Apoptosis, Cell Autophagy, Cell Confluence, Cell Count, Cell Morphology, Cell Proliferation, Cell Transfection, Lipid Droplet, Live/Dead Cell Viability, and Ring Region.
  • Versatility and Precision Achieve high precision in your life science research across diverse applications with automated processes and detailed measurement capabilities.
  • This package also includes a 12-month subscription to a Standard Success Plan from Media Cybernetics, providing ongoing technical assistance and software onboarding so you can focus on the work that truly matters.
Specifications
Condenser Type N.A.0.90/0.13 Swing In/Out Condenser
Camera Type Moticam S6
Eyepiece N-WF 10X/22 mm
Head Style Trinocular, Tilting
Lighting Type Halogen/LED
Magnification Power 2X, 4X, 10X, 20X, 40X
Model PA43BIO (Pathology)
Objective Types Plan UC
Product Type Trinocular Pathology Microscope
Nosepiece 5-position revolving nosepiece
Voltage 110/240 V