Learn More
Thermo Scientific™ CLINIConic™ Rotor Adapters
Catalog No. 75003702 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
10 mL Round-bottom Tube
5 mL Round-bottom or Blood Collection Tube
7 mL Round-bottom or Blood Collection Tube
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75-003-702 10 mL Round-bottom Tube
11-172-595 5 mL Round-bottom or Blood Collection Tube
11-172-596 7 mL Round-bottom or Blood Collection Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 75-003-702 Supplier Thermo Scientific™ Supplier No. 75003702
Add to Cart
Add to Cart
Incorporate these rotor adapters with the CLINIConic fixed-angle rotor.
Incorporate Thermo Scientific™ CLINIConic™ Rotor Adapters with the CLINIConic fixed-angle rotor.
Ordering Information:
CLINIConic fixed-angle rotor body (75-003-623) is available separately. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Tube Adapter
Capacity 10 mL Round-bottom Tube