Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cliniqa™ LiniCAL™ Calibration Verifiers

Catalog No. 2210026 Shop All CLINIQA Products
Change view
Click to view available options
Quantity:
5 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-100-26 5 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-100-26 Supplier Cliniqa™ Supplier No. 98210

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Objectively verify instrument calibration and assess linearity for specified analytes

  • True liquid, human serum based
  • Intended for use in the clinical laboratory to assess reagent/analyzer performance
  • Designed to challenge the instrument's dynamic ranges of all specified analytes
  • InterQC™ peer data and linearity verification program access included free

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes High Sensitivity C-Reactive Protein (hsCRP)
Type hsCRP
Intended Use Intended for use in the clinical laboratory to assess reagent or analyzer performance
Quantity 5 x 1.5 mL
Stability 14 Days Open Vial
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.