Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cliniqa™ Liquid QC™ Cardiac Marker Control Complete

Catalog No. 22100101 Shop All CLINIQA Products
Change view
Click to view available options
Quantity:
6 x 3 mL
2 x 3 x 3 mL
Level:
1
1, 2, 3
2
3
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Level
22-100-101 6 x 3 mL 2
22-100-100 6 x 3 mL 1
22-100-102 6 x 3 mL 3
22-100-103 2 x 3 x 3 mL 1, 2, 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Human source material. Treat as potentially infectious
Catalog No. 22-100-101 Supplier Cliniqa™ Supplier No. 91482
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Human source material. Treat as potentially infectious

Assayed quality control material for cardiac markers

  • Liquid control prepared from human plasma, human serum and human proteins
  • Assists in monitoring of analytical systems within clinical range
  • Assayed quality control material for CK-MB, d-Dimer, hsCRP, Myoglobin, Troponin I, Troponin T and NT-proBNP
  • Needs no reconstitution or frozen storage

Certifications

CE marked

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Type Cardiac Marker Control
Intended Use Assayed quality control material for cardiac markers
Level 2
Quantity 6 x 3 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.