Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cliniqa™ Liquid QC™ Cardiac Marker Control Complete
Shop All CLINIQA Products
Click to view available options
Quantity:
6 x 3 mL
2 x 3 x 3 mL
Level:
1
1, 2, 3
2
3
Description
- Liquid control prepared from human plasma, human serum and human proteins
- Assists in monitoring of analytical systems within clinical range
- Assayed quality control material for CK-MB, d-Dimer, hsCRP, Myoglobin, Troponin I, Troponin T and NT-proBNP
- Needs no reconstitution or frozen storage
Certifications
CE marked
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 8°C |
| Type | Cardiac Marker Control |
| Intended Use | Assayed quality control material for cardiac markers |
| Level | 1, 2, 3 |
| Quantity | 2 x 3 x 3 mL |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction