Learn More
Description
- Human serum based lipid control
- Assists in monitoring of analytical systems within clinical range
- Assayed quality control material for Apolipoprotein A-1, Apolipoprotein B, Cholesterol (Total), High Density Lipoprotein, Low Density Lipoprotein, Triglycerides
- Needs no reconstitution or frozen storage
- Liquid stable, two levels
Certifications
CE marked
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Apolipoprotein A1, Apolipoprotein B, Cholesterol, HDL, LDL, TG |
| Type | Lipid Control |
| Intended Use | Monitoring test procedures used for Lipid analysis |
| Level | 1 |
| Quantity | 6 x 3 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.