Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cliniqa™ Liquid QC™ Lipid Control

Catalog No. 2210052 Shop All CLINIQA Products
Change view
Click to view available options
Level:
1
2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22-100-52 2
22-100-51 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Human source material. Treat as potentially infectious
Catalog No. 22-100-52 Supplier Cliniqa™ Supplier No. 91502S

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Human source material. Treat as potentially infectious

Assayed quality control material for lipids

  • Human serum based lipid control
  • Assists in monitoring of analytical systems within clinical range
  • Assayed quality control material for Apolipoprotein A-1, Apolipoprotein B, Cholesterol (Total), High Density Lipoprotein, Low Density Lipoprotein, Triglycerides
  • Needs no reconstitution or frozen storage
  • Liquid stable, two levels

Certifications

CE marked

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Detectable Analytes Apolipoprotein A1, Apolipoprotein B, Cholesterol, HDL, LDL, TG
Type Lipid Control
Intended Use Monitoring test procedures used for Lipid analysis
Level 2
Quantity 6 x 3 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.