Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cliniqa™ Liquid QC™ Urinalysis Controls

Catalog No. 2210043 Shop All CLINIQA Products
Change view
Click to view available options
Quantity:
5 x 5 mL
2 x 25 mL
3 x 15 mL
4 x 120 mL
Type:
DipScopics Liquid QC Urinalysis Controls
DipStrip Liquid QC Urinalysis Controls
DropStrip Liquid QC Urinalysis Controls
MicroStrip Liquid QC Urinalysis Controls
For Use With (Equipment):
Dipscopics
Dipstrip
Dropstrip
Microstrip
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type For Use With (Equipment)
22-100-43 4 x 120 mL DipScopics Liquid QC Urinalysis Controls Dipscopics
22-100-40 3 x 15 mL DipStrip Liquid QC Urinalysis Controls Dipstrip
22-100-41 2 x 25 mL DropStrip Liquid QC Urinalysis Controls Dropstrip
22-100-42 5 x 5 mL MicroStrip Liquid QC Urinalysis Controls Microstrip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-100-43 Supplier Cliniqa™ Supplier No. 97204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offered in four convenient bi-level kit configurations: DipStrip, DropStrip, MicroStrip and DipScopics

  • For manual and automated test methodologies
  • For use as assayed quality control materials for Glucose, Bilirubin, Ketones, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes, hCG, Microalbumin, Creatinine, and Galactose
  • DipScopics™ also contains red blood cells, white blood cells, and crystals for microscopic evaluation of urine sediment

Specifications

Certifications/Compliance ISO 9000 13485
Content And Storage 2°C to 8°C
For Use With (Equipment) Dipscopics
Type DipScopics Liquid QC Urinalysis Controls
Intended Use In-vitro diagnostic use, use as assayed quality control materials for Glucose, Bilirubin, Ketones, specific gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes, hCG, Microalbumin, Creatinine, and Galactose
Level 2
Quantity 4 x 120 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.