Learn More
Siemens Healthineers Clinitek Status™ Connector USA WIFI
Catalog No. 23313000 Shop All Siemens Healthineers Products
Change view
Click to view available options
For Use With (Equipment):
Clinitek Status+ Analyzer
Catalog No. For Use With (Equipment)
23-313-000 Clinitek Status+ Analyzer
1 options
This item is not returnable. View return policy
Catalog No. 23-313-000 Supplier Siemens Healthineers Supplier No. 11537760
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Clinitek Status™ Connector USA WIFI is an LIS Connector base for use with the Clinitek Status™ instrument.
The connector includes the following key features:
  • Centralized control
  • Communication with an LIS
  • Bar-code scan to easily enter data
  • Operator management and security
  • Password protection
  • Improved data management
  • Quality control (QC) lockout
  • QC reminder
  • Communication through the Ethernet or a wireless connection
  • Recall patient and QC records
  • Copy Setup configuration using a USB memory stick
  • Customized urine colors
TRUSTED_SUSTAINABILITY
Specifications
Product Type Connector
For Use With (Equipment) Clinitek Status+ Analyzer
Sterility Non-sterile
WARNING: Reproductive Harm - www.P65Warnings.ca.gov