Learn More
Shockwatch Div Mri Clinitemp Temperature Monitor
SDP
Catalog No. NC2598433
Encompass_Preferred
Clinitemp features a continuous display of core-adjusted temperature as indicated by a bright line, which continuously moves across the temperature range. Readings are made at the center of the line. They are utilized throughout the surgical suite anesthesia to PACU monitoring.•84-106°F / 29 – 41°C•Easy to Read•Cost Effective•Disposable •Non – Invasive•Latex Free•Adhesive-backed paper•Made in USA•50 pieces / dispenser
Catalog No. NC2598433 Supplier Shockwatch Div Mri Supplier No. PL31450
Clinitemp features a continuous display of core-adjusted temperature as indicated by a bright line, which continuously moves across the temperature range. Readings are made at the center of the line. They are utilized throughout the surgical suite anesthesia to PACU monitoring.•84-106°F / 29 – 41°C•Easy to Read•Cost Effective•Disposable •Non – Invasive•Latex Free•Adhesive-backed paper•Made in USA•50 pieces / dispenser