Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers CLINITEST™ Rapid COVID-19 Antigen Self-Test

Catalog No. 23312022 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
5 Tests/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-022 5 Tests/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-312-022 Supplier Siemens Healthineers Supplier No. 11556711
This item is not returnable. View return policy

The CLINITEST Rapid COVID-19 Antigen Self-Test is intended to aid in the rapid detection of SARS-CoV-2 (the virus that causes COVID-19) and provides accurate results in just 15 minutes.

Easy-to-use, Easy-to-read: No special training or equipment required. All it takes is a quick and gentle nasal swab to collect the sample, drop onto the visually-read test cassette, and interpret the results. Step-by-step instructional video is available online, along with downloadable Instructions for Use (IFU).

Accurate Results for Peace of Mind: Get the answers you need with a fast, reliable, and convenient over-the-counter COVID-19 self-test. Authorized for unsupervised self-testing use by individuals aged 14 and older (or adult-collected samples from individuals ages 2-13).

Committed to Quality: Siemens Healthineers has conducted internal assessments to confirm that the CLINITEST Rapid COVID-19 Antigen Self-Test is able to detect all current Variants of Concern, including Delta and Omicron variants.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature or refrigerated storage
CE Marker Yes
Clia Complexity Waived (EUA, OTC)
Detectable Analytes SARS-CoV-2 Protein
Detection Method Lateral Flow Chromatographic Immunoassay
Sample Type Nasal Swab
Sensitivity 86.50%
Type Covid-19 Self-Test
Includes 5 test devices, 5 sterile swabs, 5 prefilled extraction tubes with buffer solution and tips, and instructions for use
Quantity 5 Tests/Box
Specificity 99.30%
Test Time 15 minutes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.