Learn More
Apexbio Technology LLC Clinofibrate 30299-08-2 200mg
SDP
Catalog No. 5000087633
Encompass_Preferred
Clinofibrate (CAS 30299-08-2) is a small-molecule lipid-regulating agent that modulates cholesterol and triglyceride metabolism It acts by inhibiting hydroxymethylglutaryl coenzyme A reductase (HMGCR) a key enzyme in the biosynthetic pathway of cholesterol Clinofibrate demonstrates an inhibitory concentration (IC50) of 0 47 mM against HMGCR indicating its potential to suppress cholesterol synthesis It is primarily utilized in biomedical research focused on the regulation of plasma lipid levels and the investigation of therapeutic strategies for hypercholesterolemia and hypertriglyceridemia
Catalog No. 50-000-87633 Supplier Apexbio Technology LLC Supplier No. B2217200
Clinofibrate (CAS 30299-08-2) is a small-molecule lipid-regulating agent that modulates cholesterol and triglyceride metabolism It acts by inhibiting hydroxymethylglutaryl coenzyme A reductase (HMGCR) a key enzyme in the biosynthetic pathway of cholesterol Clinofibrate demonstrates an inhibitory concentration (IC50) of 0 47 mM against HMGCR indicating its potential to suppress cholesterol synthesis It is primarily utilized in biomedical research focused on the regulation of plasma lipid levels and the investigation of therapeutic strategies for hypercholesterolemia and hypertriglyceridemia