Learn More
Cayman Chemical Clinofibrate, Purity: ≥95%, CAS: 30299-08-2, 50 mg
SDP
Catalog No. 502989267
Encompass_Preferred
A hypolipidemic agent; inhibits HMG-CoA reductase (IC50 = 0.47 mM); stimulates the S-indan-1-ol dehydrogenase activity of human liver AKR 1C4; reduces serum cholesterol and triglyceride levels in rats fed a normal chow pellet diet or a semisynthetic diet containing sucrose as the only carbohydrate source at 10 and 30 mg/kg
Catalog No. 50-298-9267 Supplier Cayman Chemical Supplier No. 3110950 MG
A hypolipidemic agent; inhibits HMG-CoA reductase (IC50 = 0.47 mM); stimulates the S-indan-1-ol dehydrogenase activity of human liver AKR 1C4; reduces serum cholesterol and triglyceride levels in rats fed a normal chow pellet diet or a semisynthetic diet containing sucrose as the only carbohydrate source at 10 and 30 mg/kg