Learn More
GSC Go Science Crazy Clinometer
SCI_ED
Catalog No. S28109 Shop All GSC Products
Click to view available options
Includes:
Instructions stamped on clinometer
Catalog No. Includes
S28109 Instructions stamped on clinometer
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S28109 Supplier GSC Go Science Crazy Supplier No. CLINOMETER
Not a toy. Product intended for ages 13 and up.
A tool for determining the height or slope of a tall object.
  • Easily determine the height of objects
  • Handheld device
  • Allows for determining an angle
  • For forestry and engineering
  • Instructions stamped on clinometer
Specifications
Product Type Handheld clinometer
Includes Instructions stamped on clinometer
For Use With (Application) For determining height and inclination of object
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov