Learn More
Cayman Chemical Didymin, Purity: ≥98%, CAS: 2418540-64-2, 10 mg
SDP
Catalog No. 502934777
Encompass_Preferred
A flavonoid with diverse biological activities; reduces hydrogen peroxide-induced production of ROS and cell death and increases SOD, GPx, and CAT activity in SH-SY5Y cells differentiated into a neuronal phenotype; reduces survival of A549 and H460 lung cancer cells; inhibits tumor growth in an A549 mouse xenograft model at 6 mg/kg; reduces hepatic collagen deposition, the number of hepatic lesions, and serum levels of ALT, AST, and TNF-α in a rat model of CCL4-induced hepatic fibrosis
Catalog No. 50-293-4777 Supplier Cayman Chemical Supplier No. 2519710 MG
A flavonoid with diverse biological activities; reduces hydrogen peroxide-induced production of ROS and cell death and increases SOD, GPx, and CAT activity in SH-SY5Y cells differentiated into a neuronal phenotype; reduces survival of A549 and H460 lung cancer cells; inhibits tumor growth in an A549 mouse xenograft model at 6 mg/kg; reduces hepatic collagen deposition, the number of hepatic lesions, and serum levels of ALT, AST, and TNF-α in a rat model of CCL4-induced hepatic fibrosis