Learn More
Aviva Systems Biology Corp CLINT1 Antibody - N-terminal region (ARP62506_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62506_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLINT1. It was validated on Western Blot. . Alternate names: CLINT, ENTH, EPN4, EPNR, EPSINR, FLJ46753, KIAA0171ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-698