Learn More
Apexbio Technology LLC Clioquinol 130-26-7 1g
SDP
Catalog No. 5000089821
Encompass_Preferred
Clioquinol (130-26-7) is a small-molecule modulator functioning as a metal-ion chelator It is designed to modulate metal homeostasis thereby affecting metal-dependent cellular pathways Clioquinol exerts its biological activity primarily through chelation of metal ions disrupting iron-mediated enzymatic reactions crucial for fungal and cancer cell growth In in vitro studies Clioquinol demonstrates cytotoxic effects in diverse tumor cell lines with IC50 values generally in the low micromolar range (approximately 1 10 M depending on cell type and conditions) Based on these pharmacological properties Clioquinol holds research potential in antifungal antimicrobial and oncological applications
Catalog No. 50-000-89821 Supplier Apexbio Technology LLC Supplier No. C65281000
Clioquinol (130-26-7) is a small-molecule modulator functioning as a metal-ion chelator It is designed to modulate metal homeostasis thereby affecting metal-dependent cellular pathways Clioquinol exerts its biological activity primarily through chelation of metal ions disrupting iron-mediated enzymatic reactions crucial for fungal and cancer cell growth In in vitro studies Clioquinol demonstrates cytotoxic effects in diverse tumor cell lines with IC50 values generally in the low micromolar range (approximately 1 10 M depending on cell type and conditions) Based on these pharmacological properties Clioquinol holds research potential in antifungal antimicrobial and oncological applications