Learn More
Medchemexpress LLC HY-14603 500mg Medchemexpress, Clioquinol CAS:130-26-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14603500MG
Encompass_Preferred
Medchemexpress, HY-14603 500mg Clioquinol CAS:130-26-7 Clioquinol (Iodochlorhydroxyquin) is a topical antifungal agent with anticancer activity. Clioquinol acts as an oral antimicrobial agent for the research of diarrhea and skin infections. Antibiotic. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1621