Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clioquinol, USP, 93-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601742 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-742 25 g Amber Glass Bottle
18-601-743 100 g Amber Glass Bottle
18-601-744 500 g Amber Glass Bottle
18-601-745 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-742 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CL10825GM
Add to Cart
Add to Cart

CL108, 130-26-7, C9H5CllNO

Spectrum™ Chemical Clioquinol, USP is a topical antifungal and antibacterial medication used to treat skin infections. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 130-26-7
Molecular Formula C9H5ClINO
Molecular Weight (g/mol) 305.50
InChI Key QCDFBFJGMNKBDO-UHFFFAOYSA-N
IUPAC Name 5-chloro-7-iodoquinolin-8-ol
SMILES OC1=C2N=CC=CC2=C(Cl)C=C1I

Specifications

CAS Max % 100%
Boiling Point 350.4°C (662.72°F)
Ignition Residue 0.5%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 93-100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Clioquinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.