Learn More
Grainger John Guest Pipe Clip, 1/2 in. Tube, Plastic, White, 1 ea
Catalog No. 502767246
Encompass_Preferred
These 1/2 in. pipe clips are designed to hold pipe securely in place, providing necessary support against vibration. Each clip features a hinged cover that snaps over the pipe and can be released if adjustments are required. The base includes a center hole for secure fastening with a screw, and the design is compatible with spacers to raise the clip mounting height when needed.
  • Designed for 1/2 in. pipe
  • White plastic construction
  • Hinged cover for easy access and release
  • Base mounting hole for secure installation
  • Supports pipe and vibration reduction
  • Installation does not require special tools
  • Suitable for tight spaces
  • Maximum pressure rating of 160 psi
  • Maximum temperature rating of 200°F
Catalog No. 50-276-7246 Supplier Grainger Supplier No. 16T771
These 1/2 in. pipe clips are designed to hold pipe securely in place, providing necessary support against vibration. Each clip features a hinged cover that snaps over the pipe and can be released if adjustments are required. The base includes a center hole for secure fastening with a screw, and the design is compatible with spacers to raise the clip mounting height when needed.
  • Designed for 1/2 in. pipe
  • White plastic construction
  • Hinged cover for easy access and release
  • Base mounting hole for secure installation
  • Supports pipe and vibration reduction
  • Installation does not require special tools
  • Suitable for tight spaces
  • Maximum pressure rating of 160 psi
  • Maximum temperature rating of 200°F