Learn More
Grainger John Guest Pipe Clip, 3/4 in., Plastic, White, 1 ea
Catalog No. 502767245
Encompass_Preferred
These pipe clips are designed to hold 3/4 in. PEX pipes securely in place. The clips feature a hinged cover that snaps over the pipe and can be released if necessary. The base includes a center hole for fastening with a screw, and the design supports the use of a spacer to elevate the clip for specific installation needs.
  • Fits 3/4 in. pipe
  • Constructed from plastic and PEX
  • Maximum pressure of 160 PSI
  • Maximum temperature of 200°F
  • Hinged cover design for easy pipe release
  • Base includes hole for screw mounting
  • Compatible with optional spacer for elevated mounting
  • Supports faster installation with no special tools required
  • Compliant with ASSE 1061 and 1GC 188 standards
  • Compliant with Safe Drinking Water Act for low lead in potable and non-potable water
Catalog No. 50-276-7245 Supplier Grainger Supplier No. 16T772
These pipe clips are designed to hold 3/4 in. PEX pipes securely in place. The clips feature a hinged cover that snaps over the pipe and can be released if necessary. The base includes a center hole for fastening with a screw, and the design supports the use of a spacer to elevate the clip for specific installation needs.
  • Fits 3/4 in. pipe
  • Constructed from plastic and PEX
  • Maximum pressure of 160 PSI
  • Maximum temperature of 200°F
  • Hinged cover design for easy pipe release
  • Base includes hole for screw mounting
  • Compatible with optional spacer for elevated mounting
  • Supports faster installation with no special tools required
  • Compliant with ASSE 1061 and 1GC 188 standards
  • Compliant with Safe Drinking Water Act for low lead in potable and non-potable water