Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CLIP (86-100) | 648881-58-7 | 98.5% | 1674.10 | 1 MG
SDP

Catalog No. 5000424086
Encompass_Preferred

CLIP (86-100) is a fragment consisting of amino acids 86 to 100 of the class II-associated invariant chain peptide (CLIP). This small self-peptide is a cleavage product of the invariant chain and resides in the HLA-II antigen binding groove, playing a critical role in the assembly and transport of MHC class II alphabetaIi complexes through its interaction with the class II peptide-binding site.

  • Consists of amino acids 86 to 100 of the class II-associated invariant chain peptide
  • Plays a critical role in the assembly and transport of MHC class II alphabetaIi complexes
  • Purity of 98.49%
  • Molecular weight: 1674.10
  • White to off-white solid appearance
  • Soluble in H2O at ≥ 100 mg/mL
  • Store as powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-24086 Supplier Medchemexpress LLC Supplier No. HYP18261MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CLIP (86-100) is a fragment consisting of amino acids 86 to 100 of the class II-associated invariant chain peptide (CLIP). This small self-peptide is a cleavage product of the invariant chain and resides in the HLA-II antigen binding groove, playing a critical role in the assembly and transport of MHC class II alphabetaIi complexes through its interaction with the class II peptide-binding site.

  • Consists of amino acids 86 to 100 of the class II-associated invariant chain peptide
  • Plays a critical role in the assembly and transport of MHC class II alphabetaIi complexes
  • Purity of 98.49%
  • Molecular weight: 1674.10
  • White to off-white solid appearance
  • Soluble in H2O at ≥ 100 mg/mL
  • Store as powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.