Learn More
Fine Science Tools Micro Serrefines, Jaw: 6 x 1mm, 17 mm
SDP
Catalog No. NC9903039
Encompass
Product: Micro Serrefine - Curved/17mm/ Shape: Curved/ Jaw Dimensions: 6 x 1mm/ Clamping Pressure: Medium/ Gram Pressure: 100g/ Alloy: Stainless Steel/ Length: 17mm/ Serrated: Yes/ Unit: Each
Catalog No. NC9903039 Supplier Fine Science Tools Supplier No. 1805505
Product: Micro Serrefine - Curved/17mm/ Shape: Curved/ Jaw Dimensions: 6 x 1mm/ Clamping Pressure: Medium/ Gram Pressure: 100g/ Alloy: Stainless Steel/ Length: 17mm/ Serrated: Yes/ Unit: Each