Learn More
Chemglass Life Sciences Autoclave Bag Clip
SDP
Catalog No. 50143741
Encompass_Preferred
Autoclave Bag Clip. Open and close the clip with one hand. After autoclaving, clip holds the processed bag closed for finial disposal. The closed internal dimension of the clip is 7/16”, suitable for CLS-1192 Autoclave Bags from 12 x 24” to 24 x 36”. Holds filled autoclaved bags closed during cycle. Clip permits bag to safely vent internal pressure during autoclaving. Sold 100 per case.
Catalog No. 50-143-741 Supplier Chemglass Life Sciences Supplier No. CLS1192C001
Autoclave Bag Clip. Open and close the clip with one hand. After autoclaving, clip holds the processed bag closed for finial disposal. The closed internal dimension of the clip is 7/16”, suitable for CLS-1192 Autoclave Bags from 12 x 24” to 24 x 36”. Holds filled autoclaved bags closed during cycle. Clip permits bag to safely vent internal pressure during autoclaving. Sold 100 per case.