Learn More
New England Biolabs, Inc. CLIP-Cell™ TMR-Star – 30 nmol
SDP
Catalog No. 508112083
Encompass
CLIP-Cell TMR-Star is a photostable red fluorescent substrate that can be used to label CLIP-tag fusion proteins inside living cells, on cell surfaces or in vitro.
Catalog No. 50-811-2083 Supplier New England Biolabs, Inc. Supplier No. S9219S
CLIP-Cell TMR-Star is a photostable red fluorescent substrate that can be used to label CLIP-tag fusion proteins inside living cells, on cell surfaces or in vitro.