Learn More
Roboz Surgical Binocular Magnifying Loupes, 2.5X
SDP
Catalog No. NC1243501
Encompass
These binocular magnifying loupes easily clip on a pair of glasses (glasses not provided). The pupillary distance can be adjusted with the central adjustment knob. The double hinge design allows vertical adjustments in front of the glasses so that the optical pieces can be positioned perfectly in front of the eye. Switching between magnified and normal view is possible by flipping up the loupes.
This item is not returnable. View return policy
Catalog No. NC1243501 Supplier Roboz Surgical Supplier No. ML662
These binocular magnifying loupes easily clip on a pair of glasses (glasses not provided). The pupillary distance can be adjusted with the central adjustment knob. The double hinge design allows vertical adjustments in front of the glasses so that the optical pieces can be positioned perfectly in front of the eye. Switching between magnified and normal view is possible by flipping up the loupes.