Learn More
Electron Microscopy Sciences Clip On Magnifier Diopter 10, 102 mm Working distance, 3-1/2X magnification
SDP
Catalog No. 5025018
Encompass_Preferred
Clips on to any eyeglass frame leaving hands free. Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
Working distance: 102 mm
Magnification: 3-1/2X
Working distance: 4 in
Working distance: 102 mm
Catalog No. 50-250-18 Supplier Electron Microscopy Sciences Supplier No. 6826010
Clips on to any eyeglass frame leaving hands free. Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
Working distance: 102 mm
Magnification: 3-1/2X
Working distance: 4 in
Working distance: 102 mm