Learn More
Electron Microscopy Sciences Clip On Magnifier Diopter 3, 356 mm Working distance, 1-3/4X magnification
SDP
Catalog No. 5025015
Encompass_Preferred
Clips on to any eyeglass frame leaving hands free. Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
Working distance: 356 mm
Magnification: 1-3/4X
Working distance: 14in
Working distance: 356 mm
Catalog No. 50-250-15 Supplier Electron Microscopy Sciences Supplier No. 6826003
Clips on to any eyeglass frame leaving hands free. Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
Working distance: 356 mm
Magnification: 1-3/4X
Working distance: 14in
Working distance: 356 mm